ContactUsLinkComponent
POLYVIEW® Plus HRP-DAB (Anti-Mouse) Kit, Enzo Life Sciences
Catalog # 75841-154
Supplier:  Enzo Life Sciences
CAS Number:  
POLYVIEW® Plus HRP-DAB (Anti-Mouse) Kit, Enzo Life Sciences
Catalog # 75841-154
Supplier:  Enzo Life Sciences
Supplier Number:  KIT160-0150
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    POLYVIEW® Plus HRP-DAB (Anti-Mouse) Kit
  • Size:
    150 Tests
  • Cat. No.:
    75841-154
  • Supplier no.:
    KIT160-0150
Specifications
About this item
Complete staining kit with highest sensitivity, lowest background.
  • Highest sensitivity, complete staining kit for both IHC and ISH
  • POLYVIEW® Plus reagents' biotin free nanopolymer detection circumvents endogenous background while providing specific antigen localization
  • HIGHDEF® DAB (Brown) HRP provides high intensity color development to deliver sharp, crisp staining
  • Compatible with both manual and automated platforms
The POLYVIEW® Plus HRP-DAB (Anti-Mouse) Kit is intended for both IHC and ISH staining. The kit is a non-biotin one step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. This kit can be used with blood smears, cytosmears, and cell preparations.
Ordering information: Contains IHC/ISH Peroxidase Block, Antibody Blocking Buffer, POLYVIEW® Plus HRP (Anti-Mouse) Reagent (Ready-To-Use), HIGHDEF® DAB Chromogen, HIGHDEF® DAB Substrate Buffer, HIGHDEF® Hematoxylin, Antigen Retrieval Reagent, pH 9 (10X).