ContactUsLinkComponent
SAVIEW® Plus HRP Reagent, Enzo Life Sciences
SAVIEW® Plus HRP Reagent, Enzo Life Sciences
Catalog # 75841-132
Supplier:  Enzo Life Sciences
CAS Number:  
SAVIEW® Plus HRP Reagent, Enzo Life Sciences
Catalog # 75841-132
Supplier:  Enzo Life Sciences
Supplier Number:  ACC102-0150
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    SAVIEW® Plus HRP Reagent
  • Size:
    150 Tests
  • Cat. No.:
    75841-132
  • Supplier no.:
    ACC102-0150
Specifications
About this item
High sensitivity, low background streptavidin-based nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
  • Ready-to-use reagent
  • Streptavidin-based detection with reduced carbohydrate and biotin background
  • Suitable for both manual and automated platforms
  • Suitable for custom multiplex IHC protocols
  • The Streptavidin Blocker/Diluent further reduces background (sold separately)
  • Customize with HIGHDEF® chromogens of your choice (sold separately)
SAVIEW® Plus HRP reagent is a ready-to-use streptavidin-based nanopolymer detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the SAVIEW® Plus HRP reagent can be used for in situ hybridization detection of biotinylated DNA/RNA probes.