- Description:Azostix Urinalysis Reagent Test Strips
- Cat. No.:AM2830P
- Parameters tested:Urea Nitrogen
- Supplier no.:2830P
Azostix® Urinalysis Reagent Test Strips from Siemens Healthineers are a semi-quantitative test for estimating the amount of urea nitrogen in whole blood to provide information on renal function.
- Ready-to-use test strips
- Estimate urea nitrogen in whole blood
- Requires only one drop of blood
The procedure uses one drop of whole blood. When precise results are desired a more accurate laboratory procedure is required. The strips are for professional, in vitro diagnostic use only. Azostix® urinalysis test strips are ready to use upon removal from the bottle and are read visually.