ContactUsLinkComponent
Trays, 3-Sided, MFG Tray
Trays, 3-Sided, MFG Tray
Catalog # 89022-788
Trays, 3-Sided, MFG Tray
Catalog # 89022-788
Supplier Number:  1570085269
Specifications
  • Color:
    White
  • Description:
    Trays, 3-Sided, Movable
  • Cat. No.:
    89022-788
  • Supplier no.:
    1570085269
Specifications
About this item
3-Sided movable trays for laboratory purpose with front lip firm holding to restrict movement.
  • Autoclavable
  • 1" Lips on three sides to contain spills