GeneRuler™ Biotinylated DNA Ladder, KPL INC
Catalog # 95059-388
Supplier: KPL
CAS Number:
Specifications
- Description:Biotinylated DNA Ladder
- Cat. No.:95059-388
- Lanes:20-50
- Supplier no.:600-0008
Specifications
About this item
The GeneRuler™ Biotinylated DNA Ladder is a convenient prelabeled molecular weight marker ideal for use on southern blots.
- 21 distinct bands are visible from 100 bp-10 kb
- Bright bands at 3000 bp, 1031 bp, and 500 bp serve as reference for easy orientation
The ladder contains a mixture of labeled and unlabeled DNA which migrate uniformly and will not interfere or cross hybridize with the target.