Mini Block Heater, Greiner Bio-One
Catalog # 10818-597
Supplier: Greiner Bio-One
Specifications
- Temperature range:Ambient 5...100 °C
- Description:Mini Block Heater
- Cat. No.:10818-597
Specifications
About this item
The digital block heater has an extremely compact design and provides excellent temperature uniformity and accuracy for heating of tubes from 0.2 up to 50 mL.
- Compact design
- High grade aluminum block construction
- Broad temperature range
- Multiple block options
The Mini Block Heater is intended only for indoor use and as a general laboratory tube heating device (dry bath).
Ordering information: Seven accessory blocks are available for use with a wide range of tubes (sold separately).