ContactUsLinkComponent
LKV Agar, Reducible, Hardy Diagnostics
LKV Agar, Reducible, Hardy Diagnostics
Catalog # 75846-312
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
LKV Agar, Reducible, Hardy Diagnostics
Catalog # 75846-312
Supplier:  Hardy Diagnostics
Supplier Number:  A60
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    LKV Agar, HDx
  • Size:
    15x100 mm plate
  • Cat. No.:
    75846-312
  • Supplier no.:
    A60
Specifications
About this item
LKV (Laked Sheep Blood, Kanamycin, Vancomycin) Agar, reducible, for anaerobes.
  • Selective isolation of gram negative anaerobic bacteria
  • Laked Sheep Blood, Kanamycin and Vancomycin agar
  • Reducible, for anaerobes
(Brucella Agar with Laked Sheep Blood, Kanamycin, Vancomycin, Hemin and Vitamin K) For the selective isolation of gram negative anaerobic bacteria, especially Bacteroides spp.