Pig Whole IgG IgG Isotype Control (Biotin)
Catalog # RL014-0602
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Control
- Antigen Symbol:Whole IgG
- Conjugation:Biotin
- Host:Pig
- Isotype:IgG
- Size:1 mg
- Antibody Fragment:Whole
- Cat. No.:RL014-0602
- Supplier no.:014-0602