Y-27632 dihydrochloride ≥95% (by HPLC), Calbiochem®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Y-27632 2HCl, (R)-(+)-trans-4-(1-Aminoethyl)-N-(4-Pyridyl)cyclohexanecarboxamide dihydrochloride, Rock inhibitor y-27632, trans-4-((R)-1-Aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide dihydrochloride
CAS Number:129830-38-2 | 146986-50-7 (free base)
:146986-50-7
Formula:C₁₄H₂₁N₃O·2HCl
MDL Number:MFCD03490488
MW:320.26 g/mol
:Packaged under inert gas.
:GHS07
Storage Temperature:Freezer
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specification Test Results
- Primary Target:p160 Rho-associated protein kinases (ROCK)
- Primary Target Ki:140 nM for p160 Rho-associated protein kinases (ROCK)
- Purity:≥95% by HPLC
Specification Test Results
Frequently Bought Together