ContactUsLinkComponent
Difco™ Sorbitol MacConkey Agar, BD Life Sciences
Difco™ Sorbitol MacConkey Agar, BD Life Sciences
Catalog # 90004-004
Supplier:  BD
CAS Number:  
undefined
Difco™ Sorbitol MacConkey Agar, BD Life Sciences
Catalog # 90004-004
Supplier:  BD
Supplier Number:  279100
CAS Number:  
Specifications
  • Description:
    Sorbitol MacConkey Agar
  • Size:
    500 g
  • Cat. No.:
    90004-004
  • Supplier no.:
    279100
Specifications
About this item
Sorbitol MacConkey Agar is a partially selective differential medium for the isolation of E
  • Pink colony formation
  • Peptones provide essential nutrients
  • Agar is the solidifying agent
Sorbitol-fermenting organisms produce pink colonies.
coli O157:H7 from clinical, veterinary, food, and environmental sources.