Difco™ Malonate Broth, BD Diagnostics
Catalog # 90003-110
Supplier: BD
CAS Number:
Specifications
- Description:Malonate Broth
- Size:500 g
- Cat. No.:90003-110
- Supplier no.:239520
Specifications
About this item
Malonate Broth is used for differentiating Enterobacter from Escherichia based on malonate utilization.
- Contains ammonium sulfate as a sole source of nitrogen
- Dipotassium phosphate and monopotassium phosphate provide buffering capability
- Sodium chloride maintains osmotic equilibrium
- Bromthymol blue is a pH indicator
Malonate Broth is described for differentiating Enterobacteriaceae in food and dairy products, and is suitable for organisms that would otherwise fail on the unsupplemented Leifson medium.