Difco™ Mitis Salivarius Agar, BD
Supplier: BD
Certificates
About this item
Mitis Salivarius Agar is used with Tellurite Solution 1% in isolating Streptococcus mitis.
- Peptones as a source of carbon, nitrogen, and nutrients
- Dextrose and saccharose provide carbohydrate sources
- Crystal violet and potassium tellurite inhibit bacterial growth
- Tryptan blue gives colonies a blue color
- Agar is the solidifying agent
S. mitis and S. salivarius are known as viridans streptococci, and these organisms play a role in cariogenesis and infective endocarditis and cause an increasing number of bacteremias.
Salivarius, and enterococci, particularly from grossly contaminated specimens.
Frequently Bought Together