ContactUsLinkComponent
Difco™ NIH Thioglycollate Broth, BD Life Sciences
Difco™ NIH Thioglycollate Broth, BD Life Sciences
Catalog # 90001-880
Supplier:  BD
CAS Number:  
undefined
Difco™ NIH Thioglycollate Broth, BD Life Sciences
Catalog # 90001-880
Supplier:  BD
Supplier Number:  225710
CAS Number:  
Specifications
  • Description:
    NIH Thioglycollate Broth
  • Size:
    500 g
  • Cat. No.:
    90001-880
  • Supplier no.:
    225710
Specifications
About this item
NIH Thioglycollate Broth is used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes, and microaerophiles.
  • Anaerobic incubation recommended
  • Used in same sterility procedures as FTM
  • Meets requirements of the USP growth promotion test
NIH Thioglycollate Broth is Fluid Thioglycollate Medium without the agar or indicator components.