Order Entry
United States
ContactUsLinkComponent
Difco™ MacConkey Broth, BD
Difco™ MacConkey Broth, BD
Catalog # 90001-042
Supplier:  BD
CAS Number:  
undefined
Difco™ MacConkey Broth, BD
Catalog # 90001-042
Supplier:  BD
Supplier Number:  220100
CAS Number:  

Specifications

  • Description:
    MacConkey Broth
  • Size:
    500 g
  • Cat. No.:
    90001-042
  • Supplier no.:
    220100

Specifications

About this item

MacConkey Broth is used for the detection of coliform organisms in milk and water.

  • Peptone provides amino acids and other growth factors
  • Lactose is a carbon energy source
  • Oxgall inhibits the growth of gram-positive organisms
  • Bromcresol purple is the indicator

MacConkey Broth is a modification of the original bile salt broth recommended by MacConkey that contained 0.5% sodium taurocholate and litmus as an indicator.