ContactUsLinkComponent
Difco™ Malt Extract Agar, BD Diagnostics
Difco™ Malt Extract Agar, BD Diagnostics
Catalog # 90000-120
Supplier:  BD
CAS Number:  
undefined
Difco™ Malt Extract Agar, BD Diagnostics
Catalog # 90000-120
Supplier:  BD
Supplier Number:  211220
CAS Number:  
Specifications
  • Description:
    Malt Extract Agar
  • Size:
    500 g
  • Cat. No.:
    90000-120
  • Supplier no.:
    211220
Specifications
About this item
Malt Extract Agar is used for isolating, cultivating, and enumerating yeasts and molds.
  • Maltose is an energy source
  • Dextrin and glycerol are sources of carbon
  • Peptone provides a nitrogen source
  • Agar is the solidifying agent
The acidic pH of Malt Extract Agar allows for the optimal growth of molds and yeasts while restricting bacterial growth.