Order Entry
United States
ContactUsLinkComponent
Microbore Extention Set, Braun
Microbore Extention Set, Braun
Catalog # 10753-942
Supplier:  B Braun
Microbore Extention Set, Braun
Catalog # 10753-942
Supplier:  B Braun
Supplier Number:  V5450

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Microbore Extension Set, Priming Volume of 0.7 mL
  • Cat. No.:
    10753-942
  • Supplier no.:
    V5450

Specifications

About this item

Microbore extension set with proximal and distal male luer locks.

  • Removable slide clamp
  • 1.016 mm (0.04") I.D. x 79 cm (31") Length
  • Latex-free