ContactUsLinkComponent
BBL™ Lim Broth (Todd Hewitt with CNA) For Group B Streptococcus, BD Diagnostics
BBL™ Lim Broth (Todd Hewitt with CNA) For Group B Streptococcus, BD Diagnostics
Catalog # 90004-686
Supplier:  BD
CAS Number:  
BBL™ Lim Broth (Todd Hewitt with CNA) For Group B Streptococcus, BD Diagnostics
Catalog # 90004-686
Supplier:  BD
Supplier Number:  296266
CAS Number:  
Specifications
  • Description:
    Lim Broth (Todd Hewitt with CNA)
  • Size:
    5 mL Tubes
  • Cat. No.:
    90004-686
  • Supplier no.:
    296266
Specifications
About this item
Lim Broth is used for the selective enrichment of group B streptococci (Streptococcus agalactiae).
  • Peptones, dextrose, and salts provide essential nutrients
  • Yeast extract is a source of B-complex vitamins
  • Dextrose stimulates hemolysin production
  • Nalidixic acid and colistin suppress growth of gram-negative bacteria
Lim Broth is used for the selective enrichment of group B streptococci (Streptococcus agalactiae), especially from genital specimens.