ContactUsLinkComponent
BBL™ Dermatophyte Test Medium (DTM) Modified with Chloramphenicol Slants, BD Diagnostics
BBL™ Dermatophyte Test Medium (DTM) Modified with Chloramphenicol Slants, BD Diagnostics
Catalog # 90005-248
Supplier:  BD
CAS Number:  
BBL™ Dermatophyte Test Medium (DTM) Modified with Chloramphenicol Slants, BD Diagnostics
Catalog # 90005-248
Supplier:  BD
Supplier Number:  299701
CAS Number:  
Specifications
  • Description:
    Dermatophyte Test Medium, Modified with Chloramphenicol
  • Size:
    20 x 116 mm Tubes
  • Cat. No.:
    90005-248
  • Supplier no.:
    299701
Specifications
About this item
Dermatophyte Test Medium (DTM), Modified with Chloramphenicol is a selective and differential medium used for the detection and presumptive identification of dermatophytes from clinical and veterinary specimens.
  • Soybean meal digest provides nitrogen
  • Dextrose is a source of energy
  • Phenol red indicates rise in pH of the medium
  • Antimicrobial agents inhibit bacterial growth
Dermatophyte Test Medium (DTM) is a selective and differential medium used for the detection and presumptive identification of dermatophytes from clinical specimens. Because of the unavailability of one of the inhibitory agents, chlortetracycline, Dermatophyte Test Medium (DTM), Modified with Chloramphenicol is recommended as a substitute for the original DTM formulation.