BHI (Brain Heart Infusion) Broth, Teknova
76418-032
: Teknova
:
- Description:BHI broth
- Cat. No.:76418-032
- Sterility:Sterile
- pH:7.4
- Volume:500 ml
Brain Heart Infusion (BHI) Broth is a general-purpose medium suitable for the cultivation of a wide variety of organisms, including bacteria, yeasts and molds.