To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:DCA Vantage® Analyzer
- Electrical:100 to 240 VAC, 50/60 Hz
- Weight:
3.88 kg - Cat. No.:89187-778
- Supplier no.:5075US
DCA Vantage® Analyzer for Diabetes Control from Siemens Healthcare Diagnostics is a point-of-care system that helps you monitor glycemic control and detect early kidney disease.
- Fast, actionable test results
- 1µL finger stick without fasting
- HbA1c results in 6 minutes
- Random urine sample provides A:C ratio in 7 minutes
- Requires no sample or reagent preparation prior to testing
Monitor glycemic control and diabetes complications using an analyzer designed to make consultations easier
Self-contained cartridges facilitate easy, walkaway operation after sample loading. No sample or reagent preparation required. Bar-code scanner for safer and faster patient/operator ID entry. Review results on-screen or generate a hard copy report to minimize transcription errors in the office. Convenient local storage of up to 4,000 onboard records with powerful sorting capabilities. Automatically upload results to a PC via a USB flash drive to reduce manual logging and save time.
Minimal maintenance requirements with automatic reminders to alert you when maintenance is due. Customizable security access modes support up to 1,000 operators, protect patient information, and prevent operation by unauthorized users. POCT1-A2 communication protocol streamlines data transfer for easy connectivity and fast, two-way communication to LIS/HIS, RAPIDComm® System, or other third-party POC data management systems. Automatically upload results and QC information to LIS/HIS to reduce manual logging and save time. Take testing oversight to the next level with RAPIDComm Data Management System to remotely manage multiple analyzers and operators to enhance compliance and improve risk management. POC coordinators can standardize test procedures, enforce QC protocols, control access privileges, define operator recertification requirements, and more. Customized alerts, reports, and audit trails ensure peace of mind and simplify accreditation and inspections.
Fast, actionable test results enable you to determine the effectiveness of a treatment plan, make therapeutic adjustments with confidence, and be more certain whether patients are complying with your recommendations. HbA1c from a small (1 µL) whole blood sample in 6 minutes. Flexible reporting of HbA1c% (NGSP, JDS, and Mono-S units) and IFCC (mmol/mol). HbA1c patient trending graphs can be viewed or printed. Albumin, Creatinine, and Albumin-to-Creatinine (A:C) ratio from a urine specimen in 7 minutes to report a quantitative protein status with automatic creatinine adjustment. Onboard GFR calculator indexes kidney function.
Additional information
Dimensions: 9.0 (h) x 11.5 (w) x 10.5 (d) inches 25.4 (h) x 28.7 (w) x 27.7 (d) cm
Weight: 3.88 kg (9.0 lb)
Power Requirements: 100 to 240 VAC; 50/60 Hz