Helium or Argon Gas Regulator, PerkinElmer
Catalog # 10138-306
Supplier: PerkinElmer
Specifications
- Description:Helium or Argon Gas Regulator
- Environmentally Preferable:
- Cat. No.:10138-306
- Gas:Helium/argon
- Supplier no.:N2410172
Specifications
About this item
This two-stage gas regulator has a brass body, stainless steel diaphragm and constant delivery pressure regardless of variations in cylinder pressure.
- Maximum inlet pressure is 3000 psi
- CGA fitting size is 580
An elemental analyzer uses several gases. During oxygen analysis, pyrolysis of the sample takes place in an environment of helium/hydrogen or argon/hydrogen. Nitrogen, air, or argon is required to drive the valves in the PerkinElmer 2400 and PerkinElmer 2410.
Ordering information: A Swagelok™ 1/4" npt to 1/8" fitting (Model 09903471) is included. To maintain proper pressure, leak-free connections, and remove moisture, oil, and dust from the gases, PerkinElmer regulators, pneumatic connections, driers, and traps are available.