Easy-Breathe SMS Labcoats, ValuMax
Supplier: Valumax International, Inc.
Certificates
About this item
Durable medium-weight unisex knee length lab coat for protection and comfort with SMS material.
- Fluid resistant, breathable and fire retardant
- Autoclavable and washable up to 5 times
- 3 Pockets (2 hip pockets; 1 breast pocket)
- Snap front, knit cuffs and collar
Specifications
- Catalog No:
- 89097-664
- 10119-928
- Garment application:
- Laboratory
- Laboratory
- Risk area:
- Rain (waterproof), Flame-retardant
- Rain (waterproof), Flame-retardant
- Gender:
- Unisex
- Unisex
- Closure type:
- With Press-Stud Fastening
- With Press-Stud Fastening
- Fabric:
- SMS
- SMS