Specifications
- Description:Ward's® Distinguishing Characteristics of Acids and Bases Lab Activity
- No. of lab groups:10
- Cat. No.:470023-816
- Includes:Teacher's Guide with SDS, Student Copy Master
- Supplier no.:WARD470023-816
Specifications
About this item
Explore the Distinctly Different Properties of Acids and Bases
- Introduce students to properties of acids and bases
- Students identify solutions using empirical data
Students will test the included solutions for pH, conductivity, reactivity, and more. They then use this data to identify the chemical characteristics common to acidic, neutral, and alkaline solutions.