Tak Pak® 712™ Accelerator, Loctite®, Henkel
Supplier: Loctite
Certificates
About this item
LOCTITE® SF 712™ Accelerator increases cure speed of Loctite instant adhesive
- Isopropanol base
- Excellent for cosmetic applications, for clear and transparent adhesive beads or fillets
- Suitable for securing wires or coils to PCBs
Typical applications include securing wires or coils to PCBs, tamper-proofing adjustable components, mounting stand-offs, edge guides and board stiffeners.
It is suited for post-application on cyanoacrylate
Frequently Bought Together