To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:beta-Actin
- Antigen Symbol:beta-Actin
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Antigen Synonyms:b actin|beta Actin
- Storage Buffer:Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:Antibody can be stored at 4 °C for three months and at –20 °C for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/ml
- Shipping Temperature:2-8°C
- Immunogen:Rabbit polyclonal beta-actin antibody was raised against a 16 amino acid peptide from near the amino-terminus of human beta-actin (Genbank accession No. AAH02409).
- Purification:Affinity chromatography Purified via peptide column.
- Cat. No.:10177-872
- Supplier no.:PK-AB718-3779
Rabbit Anti-Human/Mouse/Rat beta-Actin (NT)
b-actin antibody can be used for the detection of b-actin by Western blot at 1 – 2 µg/ml. (Optimal dilution should be determined by user.) Antibody can also be used for immunohistochemistry and ELISA and might be suited for other applications not tested s
Type: Primary
Antigen: beta-Actin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat