M-MLV Reverse Transcriptase Buffer Pack, Promega
Catalog # PAM5313
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:M-MLV Reverse Transcriptase Buffer Pack
- Size:2 x 1ml
- Cat. No.:PAM5313
- Supplier no.:M5313
Specifications
About this item
M-MLV RT is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb).
- RNA-Dependent DNA Polymerase for cDNA Synthesis
- Enzyme supplied at a concentration of 200u/microl
Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb). The enzyme is a product of the pol gene of M-MLV and consists of a single subunit with a molecular weight of 71kDa. The RNase H activity of M-MLV RT is weaker than that of the commonly used Avian Myeloblastosis Virus (AMV) reverse transcriptase.
Caution: Storage temperature -30°C to -10°C