ContactUsLinkComponent
T4 DNA Ligase 10X Buffer, Promega
T4 DNA Ligase 10X Buffer, Promega
Catalog # PAC1263
CAS Number:  
undefined
T4 DNA Ligase 10X Buffer, Promega
Catalog # PAC1263
Supplier Number:  C1263
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    T4 DNA Ligase 10X Buffer
  • Size:
    3×500 µl
  • Cat. No.:
    PAC1263
  • Supplier no.:
    C1263
Specifications
About this item
For ligation of DNA ends.
  • Joins Two DNA Strands with Cohesive or Blunt Ends
  • Use with 5´, 3´ or blunt-ended dsDNA (e.g., inserts and vectors)
  • Qualified for blue/white cloning, providing a higher level of quality control for enzymes used in cloning applications
  • Can catalyze the joining of RNA to either a DNA or RNA strand in a duplex molecule
T4 DNA Ligase catalyzes the joining of two strands of DNA between the 5'-phosphate and the 3'-hydroxyl groups of adjacent nucleotides in either a cohesive-ended or blunt-ended configuration. The enzyme has also been shown to catalyze the joining of RNA to either a DNA or RNA strand in a duplex molecule but will not join single-stranded nucleic acids.