Order Entry
United States
ContactUsLinkComponent
 
DNA Polymerase I Large (Klenow) Fragment, Promega
 
undefined
DNA Polymerase I Large (Klenow) Fragment, Promega
 

 

  • Storage Conditions:
    -30°C to -10°C
  • Enzyme Name:
    DNA Polymerase I Large (Klenow) Fragment
  • Concentration:
    5–10 units/µL
  • Tested Applications:
    Labeling in vitro, Sequencing, Mutagenesis, cDNA Synthesis,  Flushing Ends

 

 

Frequently Bought Together