Order Entry
United States
ContactUsLinkComponent
 
Exonuclease III
 
undefined
Exonuclease III
 

 

  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Exonuclease III
  • Concentration:
    150 - 200 units/µl
  • Tested Applications:
    Labeling in vitro, Sequencing, Mapping, Mutagenesis,  Duplex Shortening

 

 

Frequently Bought Together