Order Entry
United States
ContactUsLinkComponent
T4 RNA Ligase, Promega
T4 RNA Ligase, Promega
Catalog # PAM1051
CAS Number:  
undefined
T4 RNA Ligase, Promega
Catalog # PAM1051
Supplier Number:  M1051
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    10 U/µl
  • Description:
    T4 RNA Ligase
  • Size:
    500 U
  • Cat. No.:
    PAM1051
  • Supplier no.:
    M1051

Specifications

About this item

T4 RNA Ligase catalyzes the ATP-dependent ligation of single-stranded RNA or DNA onto the 5'-phosphoryl termini of single-stranded RNA or DNA.

  • Joins ssRNA or ssDNA onto the 5´-Phosphoryl End of ssRNA or ssDNA
  • Adds [5´-32P] nucleoside 3´,5´-bis (phosphate) onto single-stranded RNA
  • Can be inactivated by heating at 65°C for 15 minutes
  • Supplied at a concentration of 10 u/microl
  • For ligation, Labeling and cloning

T4 RNA Ligase catalyzes the ATP-dependent ligation of single-stranded RNA or DNA onto the 5'-phosphoryl termini of single-stranded RNA or DNA. The enzyme, purified from recombinant E. coli CA4 (RNase I-deficient), has an apparent molecular weight of 43.5kDa. T4 RNA Ligase also catalyzes the addition of [5'-32P] nucleoside 3',5'-bis (phosphate) onto single-stranded RNA.

Caution: Storage conditions –30...–10 °C