T3 RNA Polymerase (HC), 2500 U, Promega
Catalog # PAP4024
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:T3 RNA Polymerase (HC), 2,500u
- Size:2,500u
- Storage Conditions:-30°C to -10°C
- Enzyme Name:T3 RNA Polymerase
- Concentration:80 units/µL
- Tested Applications:Labeling in vitro, Transcription in vitro
- Cat. No.:PAP4024
- Supplier no.:P4024
Specifications
About this item
DNA-dependent RNA polymerase for in vitro transcription.
- DNA-Dependent RNA Polymerase Specific for T3 Promoter Sequences
- Exhibits extremely high affinity and specificity for T3 promoter sequences
- >90% pure as determined by SDS polyacrylamide gel electrophoresis
- Free of detectable levels of contaminating RNase and DNase activity (
- Supplied at a concentration of 10-20u/microl (Cat.# P2083) and 80u/µl (Cat.# P4024)
T3 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only T3 DNA or DNA cloned downstream from a T3 promoter can serve as a template for T3 RNA Polymerase-directed RNA synthesis.