Order Entry
United States
ContactUsLinkComponent
 
MojoSort™ Human NK Cell Isolation Kit, BioLegend
 :  Biolegend
 
undefined
MojoSort™ Human NK Cell Isolation Kit, BioLegend
 :  Biolegend

 

  • Species Reactivity:
    Human
  • Storage Temperature:
    2°C - 8°C

 

Frequently Bought Together