Specifications
- Pack type:Plastic ampoule
- Description:Endothelial Cell Growth Supplement, Bovine Neural Tissue
- Size:15 mg
- Cat. No.:80056-414
- Supplier no.:324845-15MG
Specifications
About this item
Native endothelial cell growth supplement from bovine neural tissue.
- Acts as a superior cloning factor for the isolation and growth of hybridoma cells
- Eliminate the need for feeder cell layers
A cell culture-medium supplement screened for contamination by bacteria, fungi, and mycoplasma. Mitogenic activity thought to be physically, chemically, and biologically distinct from that of FGF.