Membrane Filters, Nylon, Advantec
Supplier: Advantec MFS
Certificates
About this item
These membrane filters are inherently hydrophilic membranes made by impregnating a polyester web with nylon and are used for HPLC sample preparation, to sterilize biological solutions or buffers, or for vacuum degassing.
- High strength and heat resistance
- Binds proteins, DNA and RNA
- Pure, negligible organic extractables
- Maximum operating temperature: 180 °C
- Compatible with many aqueous and alcoholic solvents and solutions
Frequently Bought Together