


2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Boiling Pt:99 °C (1.013 hPa)
Melting Pt:–107 °C
Density:0.69 g/cm³ (20 °C) (lit.)
Flash Pt:–12 °C
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
Specification Test Results
- Vapor Density:3.9 (vs air)
- Assay (GC):Min. 99 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Suitability for residue analysis:Max. 5 ng/l
- Determination of P/N:≤5 ng/L
- Autoignition Temperature:745 °F
- Explosion Limit:6 %
- Vapor Pressure:41 mmHg ( 21 °C)
Specification Test Results
Related Information
Frequently Bought Together