


n-Heptane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
:100.20 g/mol
Boiling Pt:98.2…98.4 °C (1.000 hPa)
Melting Pt:–90.5 °C
Density:0.69 g/cm³ (15 °C) (lit.)
Flash Pt:6.7 %(V)
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS07
:GHS08
:GHS02
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance (color):complying
- Appearance (form):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.3850 - 1.3890
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Fluorescence (chinin) at 365 nm:Max. 1.0 ppb
- Fluorescence (chinin) at 254 nm:Max. 1.0 ppb
- Absorbance at 200 nm:Max. 0.70
- Absorbance at 210 nm:Max. 0.40
- Absorbance at 220 nm:Max. 0.10
- Absorbance at 230 nm:Max. 0.05
- Absorbance at 245 nm:Max. 0.01
- Absorbance at 260 nm:Max. 0.01
- Absorbance at 400 nm:Max. 0.01
- APHA:Max. 10
- Refractive Index:n20/D 1.387 (lit.)
- Autoignition Temperature:433 °F
Specification Test Results
Related Information
Frequently Bought Together