

Chloroform ≥99.8% stabilized, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
New Product
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Boiling Pt:61 °C (1.013 hPa)
Melting Pt:–61.3 °C
Density:1.485 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
ADR:6.1,III
:Stabilised with amylene
UN:1888
:GHS07
:GHS08
Specification Test Results
- Assay (GC):Min. 99.80 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as HCl):Max. 0.001 %
- Absorbance at 250 nm:Max. 1.00
- Absorbance at 260 nm:Max. 0.30
- Absorbance 280 nm:Max. 0.005
- IR spectrum:complying
- Vapor Density:4.1 (vs air)
- Refractive Index:n20/D 1.445 (lit.)
Specification Test Results
Related Information
Frequently Bought Together