Order Entry
United States
ContactUsLinkComponent
Elastin
Elastin
Catalog # 700011-252
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
Elastin
Catalog # 700011-252
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, amber
  • CAS No.:
    9007-58-3
  • Size:
    100 g
  • Protein/Peptide Name:
    Elastin
  • Formulation:
    Solution
  • Cat. No.:
    700011-252
  • Supplier no.:
    E1217-100GM

Specifications

About this item

Elastic protein, allows body tissues to return to their shape after contracting or stretching. Spectrum solutions utilize the highest quality raw materials.

Elastin, 10 Percent Solution, an elastic protein, allows body tissues to return to their shape after contracting or stretching. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST standard reference materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.