Order Entry
United States
ContactUsLinkComponent
Triple Sulfite Cycloserine Agar, Plates, Northeast Lab Services
Triple Sulfite Cycloserine Agar, Plates, Northeast Lab Services
Catalog # 101374-136
CAS Number:  
Triple Sulfite Cycloserine Agar, Plates, Northeast Lab Services
Catalog # 101374-136
Supplier Number:  P1658
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Size:
    15 x 100 mm
  • Cat. No.:
    101374-136
  • Fill volume:
    20 ml
  • Supplier no.:
    P1658

Specifications

About this item

Triple sulfite Cycloserine Agar is a prepared medium in a standard Petri Dish.

  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Prepared Medium
  • Size: 15 mm x 100 mm

The petri dish is 15 mm x 100 mm, and the fill volume is 20 mL.

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products with a high degree of consistency from batch to batch.