Order Entry
United States
ContactUsLinkComponent
Nutrient Agar, Plates, Northeast Lab Services
Nutrient Agar, Plates, Northeast Lab Services
  101374-050
 :  
Nutrient Agar, Plates, Northeast Lab Services
  101374-050
 :  P1491
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Media Format:
    Liquid
  • Size:
    15 x 100 mm
  • Cat. No.:
    101374-050
  • Fill volume:
    24 ml
  • Supplier no.:
    P1491

 

 

Nutrient Agar is contained in a standard Petri Dish.

  • Standard Petri Dish
  • Fill Volume: 24 mL or 20 mL
  • Prepared Medium
  • Various Sizes Available

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products with a high degree of consistency from batch to batch.