Order Entry
United States
ContactUsLinkComponent
Mueller Hinton Agar, Plates, Northeast Lab Services
Mueller Hinton Agar, Plates, Northeast Lab Services
Catalog # 101374-042
CAS Number:  
Mueller Hinton Agar, Plates, Northeast Lab Services
Catalog # 101374-042
Supplier Number:  P1470
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Size:
    15 x 100 mm
  • Environmentally Preferable:
  • Cat. No.:
    101374-042
  • Fill volume:
    24 ml
  • Supplier no.:
    P1470

Specifications

About this item

Mueller Hinton Agar is contained in a standard Petri Dish.

  • Standard Petri Dish
  • Fill Volume: 24 mL or 75 mL
  • Prepared Medium
  • Size: 15 mm x 100 mm
  • Five Percent Sheep's Blood

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products with a high degree of consistency from batch to batch.

Five percent sheep's blood is available.