ContactUsLinkComponent
Vented Paclitaxel Set with Polyethylene Lined Tubing, INTERLINK Injection Site
Vented Paclitaxel Set with Polyethylene Lined Tubing, INTERLINK Injection Site
Catalog # 10205-172
Supplier:  Baxter
Vented Paclitaxel Set with Polyethylene Lined Tubing, INTERLINK Injection Site
Catalog # 10205-172
Supplier:  Baxter
Supplier Number:  2C7557
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Vented paclitaxel set with polyethylene lined tubing, INTERLINK injection site, 42L×25.4W×27.4H cm (16.5×10×10.8")
  • Cat. No.:
    10205-172
  • Supplier no.:
    2C7557
Specifications
About this item
Vented paclitaxel set with polyethylene lined tubing, INTERLINK injection site with 0.22 µm downstream high pressure filter, non-DEHP 12" pump segment.
  • Male Luer lock adapter
  • Contains DEHP
  • Approximately 10 drops/ml
  • Fluid path is sterile, nonpyrogenic