Qualatex® XC 7J Anti-Static Latex Fingercots, QRP
Supplier: PIP
Certificates
About this item
The QRP 7J Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices.
- Packaged for ISO 5 (Class 100) applications
- Compliant with ASTM and IES-RP-CC 005.3 standards
- Average surface resistance of 5 X 10¹² ohms per square unit
- For use with Class II static sensitive devices (thresholds above 1000V)
7C pink Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices. The QRP 7C gloves are packaged for ISO 5 (Class 100) applications and are compliant with ASTM and IES-RP-CC 005.3 standards. Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2.
Certifications: ESD Safe to ANSI/ESD S20.20-2014.
Caution: This product contains natural rubber latex which may cause allergic reactions.
Frequently Bought Together