BIOTEK® 609BP Latex 9" Lightly Powdered AMBI Gloves, QRP
Supplier: Avantor
Certificates
About this item
609BP Biotek 9" Low Powder Latex Gloves have non-slip beaded cuffs for easy donning; resists roll down and are textured for enhanced wet/dry grip.
- Lightly powdered for wearer comfort
- Textured for enhanced wet/dry grip
- USDA compliant for food handling and processing
- Non-slip beaded cuff for easy donning; resists rolldown
BioTek gloves provide broad-based protection from acids, alkalis, and bases, while offering economical and comfortable protection.
Applications: Food handling, small parts handling, general lab, inspection, photo lab, painting and finishing, metal plating, assembly, paper processing and manufacturing.
BioTek gloves meet US Government 21-CFR specifications and are acceptable for use in food and pharmaceutical applications. Manufactured in FDA-registered facilities utilizing an FDA-registered 510(k) formulation. Latex comfort and fit; quality guaranteed.
Certifications: Exceeds ASTM D-3578.
Caution: This product contains natural rubber latex which may cause allergic reactions.
Frequently Bought Together