To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Media Format:Liquid
- Source:
- Species:
- Description:100X Non-Essential Amino Acids for Minimum Essential Medium Eagle, 2.8 - 3.8 pH
- Size:100 ml
- Environmentally Preferable:
- Sterility:Sterile
- pH:2.8-3.8
- Cat. No.:IC1681049
- Application:Growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium
- Supplier no.:1681049
Specifications
About this item
100× Non-essential Amino Acids for Minimum Essential Medium Eagle Contains 19 amino acids. The essential amino acids and the non-essential amino acids; L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
- Suitable for cell culture
- Mammalian Culture Media
- Osmolality: 59 - 89 mM/kg; Without L-glutamine
100× Non-essential Amino Acids for Minimum Essential Medium Eagle Contains 19 amino acids
May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium. In many cell culture applications using defined media for the in vitro cultivation of cells, the addition of supplemental nutrients and reagents is required. Additives such as antibiotics, buffers, and stains are frequently used to prevent bacterial contamination, control pH, and visibly monitor media conditions. Other supplements such as amino acids and vitamins are useful for enriched media beyond their normal concentrations. Attachment factors and transport factors are added to growth media to facilitate rapid cell propagation and enhance proper cell metabolism. Still other media supplements may be added to create conditions which will elicit specific cellular responses and effects, such as altered protein biosynthesis, accelerated antibody production and secretion, toxic response mechanisms, etc.
Formulation: L-Alanine: 890 mg/L, L-Asparagine H2O: 1500 mg/L, L-Aspartic Acid: 1330 mg/L, L-Glutamic Acid: 1470 mg/L, Glycine: 750 mg/L, L-Proline: 1150, L-Serine: 1050 mg/L.
The essential amino acids and the non-essential amino acids; L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.