Order Entry
United States
ContactUsLinkComponent
V-P Reagent B, Microbiology Reagents, Northeast Lab Services
V-P Reagent B, Microbiology Reagents, Northeast Lab Services
Catalog # 101375-000
CAS Number:  
V-P Reagent B, Microbiology Reagents, Northeast Lab Services
Catalog # 101375-000
Supplier Number:  X1225
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    V-P Reagent B (40% KOH)
  • Size:
    1 oz.
  • Cat. No.:
    101375-000
  • Cap type:
    20-400 black cap
  • Fill volume:
    25 ml
  • Supplier no.:
    X1225

Specifications

About this item

V-P Reagent B is contained in an amber bottle with a 20-400 black cap.

  • Amber Bottle
  • Black Cap
  • KOH Included
  • Fill Volume: 25 mL

The reagent contains KOH (potassium hydroxide).

It has a fill volume of 25 mL.