Order Entry
United States
ContactUsLinkComponent
D/E Neutralizing Broth, Tube, Northeast Lab Services
D/E Neutralizing Broth, Tube, Northeast Lab Services
Catalog # 101374-650
CAS Number:  
D/E Neutralizing Broth, Tube, Northeast Lab Services
Catalog # 101374-650
Supplier Number:  T1595
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    D/E Neutralizing Broth
  • Size:
    16 x 125 mm
  • Cat. No.:
    101374-650
  • Cap type:
    Screw cap
  • Fill volume:
    9.9 ml
  • Supplier no.:
    T1595

Specifications

About this item

D/E Neutralizing Broth is a prepared medium featuring a sturdy screw cap.

  • Prepared Medium
  • Glass Construction
  • Screw Cap
  • Fill Volume: 9.9 mL
  • Size: 16 x 125 mm

The tubes measure 16 x 125 mm and feature glass construction.

The broth has a fill volume of 9.9 mL.