Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Media Format:Liquid
- Description:Vogel Johnson Agar
- Size:15 mm x 100 mm
- Cat. No.:101374-170
- Fill volume:20 ml
- Supplier no.:P1675
Specifications
About this item
Vogel Johnson Agar is a prepared medium with a fill volume of 20 mL.
- Fill Volume: 20 mL
- Prepared Medium
- Standard Petri Dish
- Size: 15 mm x 100 mm
It's contained in a standard petri dish measuring 100 mm x 15 mm.
NEL's plated media operation utilizes carefully selected and validated ingredients.