Order Entry
United States
ContactUsLinkComponent
PHI-X Agar, Plates, Northeast Lab Services
PHI-X Agar, Plates, Northeast Lab Services
Catalog # 101374-066
CAS Number:  
undefined
PHI-X Agar, Plates, Northeast Lab Services
Catalog # 101374-066
Supplier Number:  P1501
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    PHI-X Agar
  • Size:
    15 mm x 100 mm
  • Cat. No.:
    101374-066
  • Fill volume:
    10 ml
  • Supplier no.:
    P1501

Specifications

About this item

Pseudomonas Isolation Agar is a prepared medium with a fill volume of 10 mL.

  • Fill Volume: 10 mL
  • Prepared Medium
  • Size: 15 mm x 100 mm
  • Standard Petri Dish

NEL produces a wide variety of agar-based media.

NEL's plated media operation uses carefully selected and validated ingredients for contaminant-free products.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.