Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Media Format:Liquid
- Description:Minimal Glucose Agar with Two Percent Bromcresol Purple Solution
- Size:15 mm x 100 mm
- Cat. No.:101374-032
- Fill volume:20 ml
- Supplier no.:P1430
Minimal Glucose Agar contains two percent Bromcresol Purple Solution.
- Two Percent Bromcresol Purple Solution
- Standard Petri Dish
- Fill Volume: 20 mL
- Size: 15 mm x 100 mm
- Prepared Medium
NEL's plated media operation utilizes carefully selected and validated ingredients.
This ensures contaminant-free products with a high degree of consistency from batch to batch.